Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prkl-1
Entrez Gene ID
177463
Gene Name
protein-coding
Gene Aliases
CELE_ZK381.5
Chromosome Location
Chr.IV: 6978742 - 6986935 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171372.3 NP_741435.2 2 - 3 457 94

View Full Product Details