Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-49
Entrez Gene ID
191860
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C10G11.4
Chromosome Location
Chr.I: 6297767 - 6299729 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059415.1 NP_491816.1 2 - 3 879 132

View Full Product Details