Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-1
Entrez Gene ID
183842
Gene Name
protein-coding
Gene Aliases
CELE_C55B7.6
Chromosome Location
Chr.I: 6482440 - 6485926 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001361754.3 NP_001348668.1 2 - 3 577 114
NM_059465.5 NP_491866.2 2 - 3 578 114
GenBank mRNA AY887907.1 - 2 - 3 594 114
AY954523.1 - 2 - 3 594 114

View Full Product Details