Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nuo-2
Entrez Gene ID
172363
Gene Name
Complex1_30kDa domain-containing protein
Gene Aliases
CELE_T10E9.7
Chromosome Location
Chr.I: 6530129 - 6531963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181996.5 NP_871796.1 3 - 4 484 94

View Full Product Details