Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xpg-1
Entrez Gene ID
186441
Gene Name
XPG (Xeroderma Pigmentosum group G) DNA repair gene homolog
Gene Aliases
CELE_F57B10.6
Chromosome Location
Chr.I: 6558685 - 6562346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059490.3 NP_491891.2 4 - 5 750 120

View Full Product Details