Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ulp-5
Entrez Gene ID
186900
Gene Name
ULP_PROTEASE domain-containing protein
Gene Aliases
CELE_K02F2.4
Chromosome Location
Chr.I: 6812660 - 6814249 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059551.3 NP_491952.1 3 - 4 352 150

View Full Product Details