Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctg-1
Entrez Gene ID
172431
Gene Name
protein-coding
Gene Aliases
CELE_H06O01.3
Chromosome Location
Chr.I: 7005443 - 7009200 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276522.3 NP_001263451.1 2 - 3 280 135
NM_001276523.3 NP_001263452.1 1 - 2 133 135
NM_059592.6 NP_491993.1 4 - 5 842 135

View Full Product Details