Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crml-1
Entrez Gene ID
172455
Gene Name
Carm_PH domain-containing protein
Gene Aliases
CELE_K07G5.1
Chromosome Location
Chr.I: 7152461 - 7156786 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322686.3 NP_001309450.1 4 - 5 857 100

View Full Product Details