Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttx-7
Entrez Gene ID
172477
Gene Name
protein-coding
Gene Aliases
CELE_F13G3.5
Chromosome Location
Chr.I: 7300999 - 7303177 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128981.6 NP_001122453.2 3 - 4 318 97
NM_001128982.6 NP_001122454.2 4 - 5 423 97
GenBank mRNA AB303038.1 - 3 - 4 318 97
AB303039.1 - 3 - 4 318 97

View Full Product Details