Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpb-2
Entrez Gene ID
172483
Gene Name
Guanine nucleotide-binding protein subunit beta-2
Gene Aliases
CELE_F52A8.2
Chromosome Location
Chr.I: 7318290 - 7321092 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059671.9 NP_492072.2 6 - 7 981 70

View Full Product Details