Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sue-1
Entrez Gene ID
184108
Gene Name
SUppressor of Elongation defect
Gene Aliases
CELE_F07A5.5
Chromosome Location
Chr.I: 7360199 - 7371534 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059682.5 NP_492083.2 2 - 3 752 112

View Full Product Details