Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spe-4
Entrez Gene ID
172498
Gene Name
Presenilin spe-4
Gene Aliases
CELE_ZK524.1
Chromosome Location
Chr.I: 7454660 - 7456953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059694.6 NP_492095.1 3 - 4 650 65
GenBank mRNA Z14066.1 - - 611 65

View Full Product Details