Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adr-1
Entrez Gene ID
172542
Gene Name
protein-coding
Gene Aliases
CELE_H15N14.1
Chromosome Location
Chr.I: 7773180 - 7777661 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059753.7 NP_492154.2 3 - 4 301 91
NM_182001.4 NP_871801.1 3 - 4 537 91

View Full Product Details