Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drh-3
Entrez Gene ID
172546
Gene Name
Dicer Related Helicase
Gene Aliases
CELE_D2005.5
Chromosome Location
Chr.I: 7820831 - 7826433 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059760.6 NP_492161.3 8 - 9 1454 91

View Full Product Details