Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rskn-2
Entrez Gene ID
172581
Gene Name
protein-coding
Gene Aliases
CELE_C54G4.1
Chromosome Location
Chr.I: 8014410 - 8022730 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047180.4 NP_001040645.1 11 - 12 2229 61
NM_001047181.3 NP_001040646.1 2 - 3 424 61
NM_001361775.4 NP_001348666.1 11 - 12 2234 61
NM_001361776.3 NP_001348667.1 11 - 12 2205 61

View Full Product Details