Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-2
Entrez Gene ID
172611
Gene Name
Nuclear pore complex protein Nup85
Gene Aliases
CELE_T01G9.4
Chromosome Location
Chr.I: 8291299 - 8293646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059855.6 NP_492256.1 2 - 3 633 107

View Full Product Details