Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cep-1
Entrez Gene ID
172616
Gene Name
protein-coding
Gene Aliases
CELE_F52B5.5
Chromosome Location
Chr.I: 8323910 - 8328859 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026307.7 NP_001021478.1 11 - 12 1720 150
NM_001383463.1 10 - 11 1394 150
GenBank mRNA AF440800.2 - 10 - 11 1400 150

View Full Product Details