Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fasn-1
Entrez Gene ID
172715
Gene Name
Carrier domain-containing protein
Gene Aliases
CELE_F32H2.5
Chromosome Location
Chr.I: 8975366 - 8986540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060016.8 NP_492417.2 2 - 3 167 96

View Full Product Details