Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mel-26
Entrez Gene ID
172737
Gene Name
protein-coding
Gene Aliases
CELE_ZK858.4
Chromosome Location
Chr.I: 9130485 - 9135759 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060048.11 NP_492449.2 5 - 6 1129 76

View Full Product Details