Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drsh-1
Entrez Gene ID
172830
Gene Name
protein-coding
Gene Aliases
CELE_F26E4.10
Chromosome Location
Chr.I: 9781324 - 9785724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128988.5 NP_001122460.2 4 - 5 1137 175
NM_060198.5 NP_492599.1 4 - 5 1135 175

View Full Product Details