Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aha-1
Entrez Gene ID
172889
Gene Name
Aryl hydrocarbon receptor nuclear translocator homolog
Gene Aliases
CELE_C25A1.11
Chromosome Location
Chr.I: 10193925 - 10197000 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264398.2 NP_001251327.1 1 - 2 55 106

View Full Product Details