Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mbtr-1
Entrez Gene ID
171624
Gene Name
protein-coding
Gene Aliases
CELE_Y48G1A.6
Chromosome Location
Chr.I: 343069 - 348424 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129070.2 NP_001122542.1 6 - 7 1646 62
NM_001129071.3 NP_001122543.1 6 - 7 1584 62
GenBank mRNA DQ904352.1 - 6 - 7 1586 62

View Full Product Details