Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tol-1
Entrez Gene ID
171635
Gene Name
TIR domain-containing protein
Gene Aliases
CELE_C07F11.1
Chromosome Location
Chr.I: 443999 - 461609 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025812.5 NP_001020983.1 13 - 14 3420 112

View Full Product Details