Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-21
Entrez Gene ID
191752
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_Y65B4BR.3
Chromosome Location
Chr.I: 550694 - 556709 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058350.5 NP_490751.1 3 - 4 829 61

View Full Product Details