Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smg-2
Entrez Gene ID
171696
Gene Name
Regulator of nonsense transcripts 1
Gene Aliases
CELE_Y48G8AL.6
Chromosome Location
Chr.I: 1176473 - 1190829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058428.6 NP_490829.1 8 - 9 2914 82

View Full Product Details