Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-10
Entrez Gene ID
171763
Gene Name
DNA-directed RNA polymerases I, II, and III subunit RPABC5
Gene Aliases
CELE_Y37E3.3
Chromosome Location
Chr.I: 2064293 - 2064686 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058522.5 NP_490923.1 1 - 2 99 91

View Full Product Details