The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306434.3 | NP_001293363.1 | 1 - 2 | 121 | 73 |
NM_058555.9 | NP_490956.1 | 2 - 3 | 278 | 73 | |
GenBank mRNA | AF300704.1 | - | 2 - 3 | 280 | 73 |