Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-17
Entrez Gene ID
171832
Gene Name
protein-coding
Gene Aliases
CELE_Y71F9B.5
Chromosome Location
Chr.I: 2706178 - 2717549 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026642.5 NP_001021813.1 9 - 10 2583 88
NM_001306438.3 NP_001293367.1 6 - 7 1347 88
NR_131370.1 6 - 7 1381 88
GenBank mRNA U63557.1 - 8 - 9 1591 88

View Full Product Details