Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snr-7
Entrez Gene ID
171834
Gene Name
Probable small nuclear ribonucleoprotein G
Gene Aliases
CELE_Y71F9B.4
Chromosome Location
Chr.I: 2726683 - 2727545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058631.7 NP_491032.1 2 - 3 149 66
GenBank mRNA AF303264.1 - 1 - 2 129 66

View Full Product Details