Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plk-2
Entrez Gene ID
171838
Gene Name
Serine/threonine-protein kinase plk-2
Gene Aliases
CELE_Y71F9B.7
Chromosome Location
Chr.I: 2755540 - 2758204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058635.8 NP_491036.1 4 - 5 1146 67
GenBank mRNA AF194964.1 - 4 - 5 1168 67

View Full Product Details