Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-49
Entrez Gene ID
187239
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_K09H9.3
Chromosome Location
Chr.I: 3137185 - 3138447 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058705.5 NP_491106.1 1 - 2 286 106

View Full Product Details