Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vbh-1
Entrez Gene ID
171888
Gene Name
protein-coding
Gene Aliases
CELE_Y54E10A.9
Chromosome Location
Chr.I: 3150341 - 3162391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026622.2 NP_001021793.1 5 - 6 844 86
NM_001383938.1 5 - 6 694 86
NM_058712.4 NP_491113.1 5 - 6 685 86

View Full Product Details