Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nol-58
Entrez Gene ID
171901
Gene Name
Nop domain-containing protein
Gene Aliases
CELE_W01B11.3
Chromosome Location
Chr.I: 3267094 - 3269122 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383952.1 2 - 3 626 88

View Full Product Details