Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnp-6
Entrez Gene ID
171922
Gene Name
protein-coding
Gene Aliases
CELE_Y47G6A.20
Chromosome Location
Chr.I: 3539554 - 3546910 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058775.7 NP_491176.1 6 - 7 999 64

View Full Product Details