Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cah-6
Entrez Gene ID
189049
Gene Name
Alpha-carbonic anhydrase domain-containing protein
Gene Aliases
CELE_T28F2.3
Chromosome Location
Chr.I: 3628246 - 3631112 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058788.3 NP_491189.1 2 - 3 358 67

View Full Product Details