Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-13
Entrez Gene ID
171943
Gene Name
Innexin
Gene Aliases
CELE_Y8G1A.2
Chromosome Location
Chr.I: 3748515 - 3753958 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058811.7 NP_491212.1 6 - 7 1026 69

View Full Product Details