Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgk-1
Entrez Gene ID
171965
Gene Name
Probable phosphoglycerate kinase
Gene Aliases
CELE_T03F1.3
Chromosome Location
Chr.I: 3867535 - 3869109 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058844.7 NP_491245.1 1 - 2 160 54

View Full Product Details