Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-53
Entrez Gene ID
171967
Gene Name
C-type LECtin
Gene Aliases
CELE_T03F1.10
Chromosome Location
Chr.I: 3871474 - 3873428 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058846.5 NP_491247.2 3 - 4 576 80

View Full Product Details