Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-89
Entrez Gene ID
171990
Gene Name
protein-coding
Gene Aliases
CELE_C09D1.1
Chromosome Location
Chr.I: 4035702 - 4090885 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025814.4 NP_001020985.1 36 - 37 20448 99
NM_001025818.3 NP_001020989.1 34 - 35 18528 99
NM_001306524.2 NP_001293453.1 2 - 3 479 99
NM_001306525.3 NP_001293454.1 2 - 3 528 99
NM_001356807.3 NP_001343717.1 35 - 35 18288 99
NM_001356808.3 NP_001343716.1 36 - 36 18729 99
NM_001356809.3 NP_001343714.1 35 - 35 20382 99
NM_001356812.3 NP_001343617.1 27 - 28 19029 99
NM_001356813.3 NP_001343719.1 26 - 27 16869 99
NM_001356814.3 NP_001343711.1 27 - 28 17310 99
NM_001356815.3 NP_001343712.1 26 - 27 18963 99
NM_001356816.3 NP_001343713.1 25 - 26 17109 99

View Full Product Details