Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cogc-5
Entrez Gene ID
172025
Gene Name
Conserved Oligomeric Golgi (COG) Component
Gene Aliases
CELE_C43E11.11
Chromosome Location
Chr.I: 4249493 - 4253887 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058938.8 NP_491339.2 4 - 5 1813 78

View Full Product Details