Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scpl-3
Entrez Gene ID
172032
Gene Name
protein-coding
Gene Aliases
CELE_Y47D9A.2
Chromosome Location
Chr.I: 4298285 - 4302245 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348610.3 NP_001335530.1 1 - 2 107 75
NM_001382905.1 3 - 4 501 75

View Full Product Details