Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-6
Entrez Gene ID
191479
Gene Name
AcetylCholine Receptor
Gene Aliases
CELE_ZK973.5
Chromosome Location
Chr.I: 4359207 - 4362548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058953.3 NP_491354.2 2 - 3 233 101

View Full Product Details