Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-27
Entrez Gene ID
182508
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_C10H11.5
Chromosome Location
Chr.I: 4724065 - 4726771 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059035.3 NP_491436.1 2 - 3 226 108

View Full Product Details