Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-28
Entrez Gene ID
172086
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C10H11.4
Chromosome Location
Chr.I: 4727280 - 4730366 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059036.6 NP_491437.1 5 - 6 1477 89

View Full Product Details