Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-38
Entrez Gene ID
172105
Gene Name
Acetylcholine receptor subunit alpha-type unc-38
Gene Aliases
CELE_F21F3.5
Chromosome Location
Chr.I: 4897620 - 4901023 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059071.7 NP_491472.1 7 - 8 1580 107
GenBank mRNA X98600.1 - - 1526 107

View Full Product Details