Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-63
Entrez Gene ID
172150
Gene Name
Acetylcholine receptor subunit alpha-type unc-63
Gene Aliases
CELE_Y110A7A.3
Chromosome Location
Chr.I: 5141673 - 5149226 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059132.5 NP_491533.2 7 - 8 1158 63
GenBank mRNA AF288374.1 - 7 - 8 1156 63

View Full Product Details