Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spd-5
Entrez Gene ID
172155
Gene Name
Spindle-defective protein 5
Gene Aliases
CELE_F56A3.4
Chromosome Location
Chr.I: 5171285 - 5176664 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059138.5 NP_491539.1 4 - 5 1420 72

View Full Product Details