Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhs-1
Entrez Gene ID
172172
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_C01G8.3
Chromosome Location
Chr.I: 5278803 - 5281526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059156.6 NP_491557.1 7 - 8 848 115

View Full Product Details