Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
coq-1
Entrez Gene ID
172190
Gene Name
COenzyme Q (ubiquinone) biosynthesis
Gene Aliases
CELE_C24A11.9
Chromosome Location
Chr.I: 5412841 - 5415629 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059187.6 NP_491588.1 3 - 4 344 102

View Full Product Details