Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpy-5
Entrez Gene ID
172197
Gene Name
Cuticle collagen dpy-5
Gene Aliases
CELE_F27C1.8
Chromosome Location
Chr.I: 5432197 - 5433051 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059194.6 NP_491595.1 1 - 1 234 62

View Full Product Details